Ammoniaklösung 20%, NORMATOM®, ultrarein für die Spurenanalytik von Metallen

Lieferant: VWR Chemicals
Gefahr

Synonyme: AmmoniakAmmoniumhydroxid

85031.230EA 275 EUR
85031.230 85031.270
Ammoniaklösung 20%, NORMATOM®, ultrarein für die Spurenanalytik von Metallen
Ammoniaklösung
Formel: NH₃
Molecular Weight: 35,05 g/mol
Dichte: 0,923 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00066650
CAS-Nummer: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8,III

For metal trace analysis in AAS or ICP

Ask for your “customized multi-element standard solution” with your exact composition using the request form

Ask for a quotation

12/31/2022

Bestellen

Spezifikation Testergebnisse

Gehalt 20 - 22%
Ag (Silber) 10 ppt
Al (Aluminium) 20 ppt
As (Arsen) 10 ppt
Au (Gold) 10 ppt
Ba (Barium) 10 ppt
Be (Beryllium) 10 ppt
Bi (Bismut) 10 ppt
Ca (Calcium) 10 ppt
Cd (Cadmium) 10 ppt
Ce (Cer) 10 ppt
Co (Cobalt) 10 ppt
Cr (Chrom) 10 ppt
Cs (Cäsium) 10 ppt
Cu (Kupfer) 10 ppt
Dy (Dysprosium) 10 ppt
Er (Erbium) 10 ppt
Eu (Europium) 10 ppt
Fe (Eisen) 10 ppt
Ga (Gallium) 10 ppt
Gd (Gadolinium) 10 ppt
Ge (Germanium) 10 ppt
Hf (Hafnium) Nur information
Hg (Quecksilber) 200 ppt
Ho (Holmium) 10 ppt
In (Indium) 10 ppt
K (Kalium) 10 ppt
La (Lanthan) 10 ppt
Li (Lithium) 10 ppt
Lu (Lutetium) 10 ppt
Mg (Magnesium) 10 ppt
Mn (Mangan) 10 ppt
Mo (Molybdän) 10 ppt
Na (Natrium) 20 ppt
Nb (Niob) 10 ppt
Nd (Neodym) 10 ppt
Ni (Nickel) 10 ppt
Pb (Blei) 10 ppt
Pd (Palladium) Nur information
Pr (Praseodym) 10 ppt
Pt (Platin) Nur information
Rb (Rubidium) 10 ppt
Re (Rhenium) Information only
Rh (Rhodium) 10 ppt
Ru (Ruthenium) Nur information
Sb (Antimon) 10 ppt
Sc (Scandium) 10 ppt
Se (Selen) Nur information
Sm (Samarium) 10 ppt
Sn (Zinn) 10 ppt
Sr (Strontium) 10 ppt
Tb (Terbium) 10 ppt
Te (Tellur) 10 ppt
Th (Thorium) 10 ppt
Ti (Titan) 10 ppt
Tl (Thallium) 10 ppt
Tm (Thulium) 10 ppt
U (Uran) 10 ppt
V (Vanadium) 10 ppt
W (Wolfram) 10 ppt
Y (Yttrium) 10 ppt
Yb (Ytterbium) 10 ppt
Zn (Zink) 10 ppt
Zr (Zirkon) 10 ppt

For metal trace analysis in AAS or ICP

Ask for your “customized multi-element standard solution” with your exact composition using the request form

Ask for a quotation

12/31/2022

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR