Salpetersäure 67%, NORMATOM®, ultrarein für die Spurenanalytik von Metallen

Lieferant: VWR Chemicals
Gefahr

83879.300EA 1600 EUR
83879.300 83879.230 83879.270 83879.290
Salpetersäure 67%, NORMATOM®, ultrarein für die Spurenanalytik von Metallen
Salpetersäure
Formel: HNO₃
Molecular Weight: 63,01 g/mol
Siedepunkt: 120,5 °C (1013 hPa)
Schmelzpunkt: –42 °C
Dichte: 1,4 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011349
CAS-Nummer: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8 (5.1),II
Merck Index: 13,06608

For metal trace analysis in AAS or ICP

Ask for your “customized multi-element standard solution” with your exact composition using the request form

Ask for a quotation

12/31/2022

Bestellen

Spezifikation Testergebnisse

Gehalt Min. 67 %
Ag (Silber) Max. 10 ppt
Al (Aluminium) Max. 20 ppt
As (Arsen) Max. 20 ppt
Au (Gold) Max. 20 ppt
B (Bor) Max. 20 ppt
Ba (Barium) Max. 10 ppt
Be (Beryllium) Max. 10 ppt
Bi (Bismut) Max. 10 ppt
Ca (Calcium) Max. 20 ppt
Cd (Cadmium) Max. 10 ppt
Ce (Cer) Max. 10 ppt
Co (Cobalt) Max. 10 ppt
Cr (Chrom) Max. 20 ppt
Cs (Cäsium) Max. 10 ppt
Cu (Kupfer) Max. 20 ppt
Dy (Dysprosium) Max. 1 ppt
Er (Erbium) Max. 1 ppt
Eu (Europium) Max. 1 ppt
Fe (Eisen) Max. 20 ppt
Ga (Gallium) Max. 10 ppt
Gd (Gadolinium) Max. 1 ppt
Ge (Germanium) Max. 10 ppt
Hf (Hafnium) Max. 10 ppt
Hg (Quecksilber) Max. 100 ppt
Ho (Holmium) Max. 1 ppt
In (Indium) Max. 1 ppt
K (Kalium) Max. 10 ppt
La (Lanthan) Max. 1 ppt
Li (Lithium) Max. 10 ppt
Lu (Lutetium) Max. 1 ppt
Mg (Magnesium) Max. 10 ppt
Mn (Mangan) Max. 10 ppt
Mo (Molybdän) Max. 10 ppt
Na (Natrium) Max. 10 ppt
Nb (Niob) Max. 1 ppt
Nd (Neodym) Max. 1 ppt
Ni (Nickel) Max. 50 ppt
Pb (Blei) Max. 10 ppt
Pd (Palladium) Max. 20 ppt
Pr (Praseodym) Max. 1 ppt
Pt (Platin) Max. 20 ppt
Rb (Rubidium) Max. 10 ppt
Re (Rhenium) Max. 10 ppt
Rh (Rhodium) Max 10 ppt
Ru (Ruthenium) Max. 20 ppt
Sb (Antimon) Max. 10 ppt
Sc (Scandium) Max. 10 ppt
Sm (Samarium) Max. 1 ppt
Sn (Zinn) Max. 20 ppt
Sr (Strontium) Max. 10 ppt
Tb (Terbium) Max. 1 ppt
Te (Tellur) Max. 1 ppt
Th (Thorium) Max. 1 ppt
Ti (Titan) Max. 10 ppt
Tl (Thallium) Max. 10 ppt
Tm (Thulium) Max. 1 ppt
U (Uran) Max. 1 ppt
V (Vanadium) Max. 10 ppt
W (Wolfram) Max. 10 ppt
Y (Yttrium) Max. 1 ppt
Yb (Ytterbium) Max. 1 ppt
Zn (Zink) Max. 20 ppt
Zr (Zirkon) Max. 10 ppt

For metal trace analysis in AAS or ICP

Ask for your “customized multi-element standard solution” with your exact composition using the request form

Ask for a quotation

12/31/2022

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR