Wasser, GPR RECTAPUR®

Lieferant: VWR Chemicals

Synonyme: WasserstoffoxidHydrogenoxid

83612.360EA 22.1 EUR
83612.360 83612.460 83612.550 83612.410 83612.400
Wasser, GPR RECTAPUR®
Wasser
Formel: H₂O
Molecular Weight: 18,02 g/mol
Siedepunkt: 100 °C (1013 hPa)
Schmelzpunkt: 0 °C
Dichte: 1 g/cm³ (4 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011332
CAS-Nummer: 7732-18-5
EINECS: 231-791-2
Merck Index: 12,10175

Bestellen

Spezifikation Testergebnisse

Acidity or alkalinity Passes test
Oxidisable substances Passes test
Conductivity (20°C) Max. 1.5 µS/cm
Evaporation residue Max. 1 ppm
Heavy metals (as Pb) Max. 0.1 ppm
Cl (Chloride) Max. 0.1 ppm
NH4 (Ammonium) Max. 0.2 ppm
NO3 (Nitrate) Max. 0.2 ppm
SO4 (Sulphate) Max. 1 ppm
Ca (Calcium) Max. 0.1 ppm
Na (Sodium) Max. 0.1 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR