Microplates, Well-Coated™
Lieferant: G-Biosciences
Well-Coated™
786-780EA 397 EUR
786-780 786-741 786-763 786-764 786-743 786-765 786-766 786-781 786-760 786-761 786-762 786-739 786-778 786-756 786-757 786-779 786-735 786-758 786-759 786-737 786-774 786-775 786-731 786-753 786-776 786-733 786-755 786-777 786-770 786-771 786-772 786-773 786-751 786-749 786-767 786-745 786-768 786-769 786-747
Microplates, Well-Coated™
Mikrotiterplatten Immunologie-Mikrotiterplatten
Pre-coated, blocked microtitre plates for screening and rapid assays.
  • Available in clear, black or white for colorimetric, chemiluminescent and fluorescent detection systems
  • 8 well strip format, supplied in convenient 96 well holder
  • Pre-blocked to save time
Protein A, G or A/G
Binds the constant (Fc) region of immunoglobulins ensuring that the antigen binding domain of the antibody is orientated away from the plate, offering maximum exposure of the binding site.
Protein L
Binds the kappa light chains of immunoglobulins without interfering with the antigen binding site; binds to all classes of immunoglobulin, including IgG, IgM, IgA, IgE and IgD. Also binds to single chain variable fragments (scFv and Fab).
Goat α-mouse IgG antibody or α-rabbit IgG antibody
Suitable for binding assays using low quantities of antibodies or antibodies that denature on direct binding to polystyrene plates.
Biotin binding
Designed to specifically bind avidin, steptavidin or Neutravidin™ conjugated molecules, including enzyme conjugates.
Nickel
Binding of 6X His tagged, expressed recombinant proteins.
Glutathione
Specific binding of GST tagged, expressed recombinant proteins.
Amine or sulfhydryl binding
Custom design ELISA plates and couple amine or sulfhydryl containing peptides, proteins, and compounds.