Protease inhibitor cocktails, ProteCEASE™
Lieferant: G-Biosciences
786-326TEA 146 EUR
786-326T 786-327 786-328 786-334 786-335 786-336 786-326
Protease inhibitor cocktails, ProteCEASE™
Zellkultur-Ergänzungen und -Zusätze
ProteCEASE™ is a dry format version of ProteaseArrest™ for large scale preparative applications and for those users who prefer reconstitution prior to use. ProteCEASE™ is a superior general protease inhibitor cocktail that is suitable for purification from mammalian, plant, bacteria and yeast samples. The cocktail contains both irreversible and reversible protease inhibitors to inhibit serine, cysteine and other proteases. EDTA is an optional component for inhibiting metalloproteases.
  • High protease inhibition: >95% in mouse pancreas lysate
  • Inhibits a wide variety of proteases, including cysteine, serine and metallo-proteases
  • Available with or without EDTA
  • Performs with a wide range of samples, including animals, plants, yeast, bacteria and fungal lysates
The EDTA-free ProteCEASE™ will maintain activity of proteins dependent on divalent cations and will not inhibit the purification of proteins with immobilised metal affinity chromatography (IMAC).
Applications include large scale preparative protein purifications, inhibition of protease activity in protein preparations, and protection of proteins from proteolysis in such applications as electrophoresis, purification, storage, assays and other applications.
  • High protease inhibition: >95% in mouse pancreas lysate
  • Inhibits a wide variety of proteases, including cysteine, serine and metallo-proteases
  • Available with or without EDTA
  • Performs with a wide range of samples, including animals, plants, yeast, bacteria and fungal lysates
Lieferumfang: ProteCEASE™ is available in two vial sizes: ProteCEASE™-50 and ProteCEASE™-100, for 50 and 100 ml of lysis buffer, respectively. ProteCEASE™-50 is available in packs of 10 or 20 vials for a total volume of either 500 ml or 1 litre. ProteCEASE™-100 is available in packs of 10 for 1 litre total volume.