Protease inhibitor cocktails, ProteaseArrest™
Lieferant: G-Biosciences
786-437EA 762 EUR
786-437 GENO786-712 786-329 786-711 733-1996
Protease inhibitor cocktails, ProteaseArrest™
Zellkultur-Ergänzungen und -Zusätze
The ProteaseArrest™ family of protease inhibitor cocktails are supplied as convenient, ready-to-use, liquid format cocktails that offer immediate >95% inhibition of harmful proteases. The 100X concentrated solution outperforms other similar protease inhibitor cocktails (including tablet formats) by offering instant inhibition, allowing delivery of optimised concentrations, demonstrating higher inhibition levels, and providing a greater ease of use. Also available as single use OneQuant™ ProteaseArrest™ aliquots.
  • Ready-to-use, broad spectrum, 100X concentrated protease inhibitor cocktail
  • High inhibition levels: 1X ProteaseArrest™ inhibits >95% of protease activities
  • Outperforms tablet cocktails
  • Excellent inhibition of protease activity in preparations of mammalian, bacteria, plant, yeast and fungal lysates
  • Protection of proteins in such applications as electrophoresis, purification, storage, assays and more
  • Suitable for small volume, analytical sample applications