Microplates, 96-Well
Lieferant: Greiner bio-one
LUMITRAC™ 600 FLUOTRAC™ LUMITRAC™ 200
736-0223EA 206 EUR
736-0223 391-3629 GREI655944_5 391-3605 738-0208 391-3615 736-0119 738-0016 736-0229 391-3610 738-0168 391-3311 738-0023 738-0021 736-0228 738-0022 738-0020
Microplates, 96-Well
Mikrotiterplatten HTS-Mikrotiterplatten
PS, without lid. A large number of different 96-well microplates are available in a wide variety of surface treatments with a choice of well profile, and with either solid or µClear® bottom. All plates are free of DNase, RNase and human DNA, and are non-pyrogenic.