384-well microplates, Falcon®
Lieferant: Corning
Falcon®
736-2044EA 817.19 EUR
736-2044 736-2043 736-2057 736-2045
384-well microplates, Falcon®
Mikrotiterplatten HTS-Mikrotiterplatten
Falcon® 384 microplates are compatible with automation systems and meet standard ANSI/SBS footprint dimensions for microplates.