Storage Plates, 96-Well, MASTERBLOCK®
Lieferant: Greiner bio-one
MASTERBLOCK®
736-0133EA 713 EUR
736-0133 736-0155 736-0156 736-0134 GREI780280_50 736-0135 736-0130 736-0131 736-0132
Storage Plates, 96-Well, MASTERBLOCK®
Mikrotiterplatten Deep-Well-Mikroplatten
PP. The 0,5 ml, 1 ml and 2 ml MASTERBLOCK® microplates are ideal for storing samples or for cultivating bacteria or yeasts.
  • Biologically inert, resistant to commonly used solvents (such as DMSO, acetonitrile or acetone) and temperature resistant (−196 °C to +121 °C)
  • Uniform external dimensions and tolerances with SBS compatible footprint
  • Elevated well walls allows them to be easily sealed with adhesive films, heat seals or CapMats
  • Alphanumeric well coding
  • Available sterile or non-sterile, and in a choice of colours