Corning® X‐LAB® systems human CD8 / CD3 selection kits

Lieferant: Corning
Corning®
734-3381EA 766.35 EUR
734-3381 734-3382
Corning® X‐LAB® systems human CD8 / CD3 selection kits
Bioreaktoren
The Corning® X-MINI® selection kits are designed for the selection of cells and cell subsets from blood and blood products using buoyancy activated cell sorting (Corning X-BACS™) technology ensuring high purity, consistency and viability of cells for downstream applications.

  • Simplified process: Easy to use
  • Single-use, column-free: Gentle on your cells
  • High purity: Achieve purities of up to 99%
  • Consistency: Reproducible results (>86%)
  • High viability (>97%)
  • Process up to 4×108 total cells

These kits are designed for the selection of cells and cell subsets from blood and blood products using buoyancy activated cell sorting (Corning X-BACS™) technology and a biotinylated antibody conjugate. Selected cells are immediately available for downstream applications such as flow cytometry and cell culture expansion.
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR