Corning® rLaminin-521 (human)
Lieferant: Corning
Corning®
734-2757EA 2374.93 EUR
734-2757 734-2755 734-2756
Corning® rLaminin-521 (human)
Zellkultur-Ergänzungen und -Zusätze
Corning® rLaminin-521 (human) is a heterotrimer composed of α5, β2, and γ1 chains expressed in a mammalian cell culture system. rLaminin-521 (human) supports long-term self-renewal of human pluripotent stem cells (hPSC), including embryonic stem cells (hESC) and induced pluripotent stem cells (iPSC) in defined and xeno-free environments. rLaminin-521 provides additional benefits, including ROCK inhibitor independent single cell expansion of PSCs, which improves hPSC culture ease and efficiency.