Spinner flasks with vertical sidearms, Corning®
Lieferant: Corning
Corning®
734-1931EA 1755.24 EUR
734-1931 734-1930 734-1933 734-1932 734-1928 734-1927 734-1926 734-1929
Spinner flasks with vertical sidearms, Corning®
Kolben Spinnerkolben
Borosilicate glass flasks are baffled to enhance agitation of contents.
  • Side baffles enhance aeration and agitation of flask contents
  • Proprietary 'stretch' impeller design ensures optimal stirring
  • Vertical sidearms permit use with sidearm probes and fittings
Can be used with direct drive motor or magnetically driven paddle assembly.
Consistent height to width ratios for greater predictability in scale up across the full range of vessel sizes.