Biologische Testschalen, 245×245 mm, quadratisch, Corning®
Lieferant: Corning
Corning®
734-1732EA 277.42 EUR
734-1732 734-1731 734-1728
Biologische Testschalen, 245×245 mm, quadratisch, Corning®
Zellkulturschalen Biologische Testschalen
Optically clear PS. For applications where cell attachment is not desired.
  • With stacking beads to aid handling
  • Supplied with vents to provide consistent gas exchange
  • Sterilized by gamma radiation
  • Certified non-pyrogenic