Amine and carboxyl coated plates, Corning® PureCoat™
Lieferant: Corning Life Sciences
Corning® PureCoat™
734-1478EA 1250.6 EUR
734-1478 734-1477 734-1472 734-1483 734-1471 734-1476 734-1475 734-1486 734-1485 734-1474 734-1484 734-1473
Amine and carboxyl coated plates, Corning® PureCoat™
Mikrotiterplatten Zellkultur-Platten Zellkulturplatten, beschichtet
PS. Corning® PureCoat™ surfaces are a novel family of chemically-defined, animal-free cell culture surfaces designed to enhance cell performance. A proprietary thin-film coating technology produces a uniform, functionalised surface that provides a highly controlled environment for cell culture applications.
  • Available as amine (positively charged) or carboxyl (negatively charged) surface chemistries
  • Optimised for culture in low serum or no serum conditions
  • Enhanced freeze-thaw recovery of cryopreserved cells
  • Improved differentiation of cells
  • Culture of many cell lines and primary cells supported, including primary neuronal cells and stem cells