Cell culture dishes, amine and carboxyl coated, Corning® PureCoat™
Lieferant: Corning
Corning® PureCoat™
734-1489EA 46.39 EUR
734-1489 734-1490 734-1482 734-1481
Cell culture dishes, amine and carboxyl coated, Corning® PureCoat™
Zellkulturschalen Zell- und Gewebekultur-Platten
Corning® PureCoat™ surfaces are a novel family of chemically-defined, animal-free cell culture surfaces designed to enhance cell performance. A proprietary thin-film coating technology produces a uniform, functionalised surface that provides a highly controlled environment for cell culture applications.
  • Available as amine (positively charged) or carboxyl (negatively charged) surface chemistries
  • Optimised for culture in low serum or no serum conditions
  • Enhanced freeze-thaw recovery of cryopreserved cells
  • Improved differentiation of cells
  • Culture of many cell lines and primary cells supported, including primary neuronal cells and stem cells