Kultursupplement mit ConA, Ratte, Corning®
Lieferant: Corning
Corning®
734-1310EA 613.42 EUR
734-1310
Kultursupplement mit ConA, Ratte, Corning®
Zellkultur-Ergänzungen und -Zusätze
T-STIM with ConA (IL-2 culture supplement), rat, is used to promote proliferation and activation of T-cells and support high-titer HIV production by leukocytes.
Media used to promote proliferation and activation (PHA or ConA Supplemented Media) of T-cells and support high-titer HIV production by leukocytes.
Tested for ability to promote proliferation of an IL-2 dependent mouse cell line (CTLL) in serum-containing medium. Filtered (0.2 µm membrane) and tested and found negative for the presence of bacteria, fungi, and mycoplasma.