Kulturmedium für Endothelzellen, Corning® BioCoat™
Lieferant: Corning
Corning® BioCoat™
734-1086EA 693.55 EUR
734-1086
Kulturmedium für Endothelzellen, Corning® BioCoat™
Zellkultur-Ergänzungen und -Zusätze
Corning® BioCoat™ Intestinal Epithelium Differentiation Environment is an integrated system designed to create in vitro intestinal models.
Lieferumfang: Kit comprises Corning® BioCoat™ fibrillar collagen cell culture inserts (24 inserts, one µm pore size in two 24-well plates), intestinal epithelium seeding basal medium (100 ml), intestinal epithelium differentiation medium (400 ml), and MITO+ serum extender (0,5 ml).