Collagen I Zellkulturschalen, Corning® BioCoat™
Lieferant: Corning
Corning® BioCoat®
734-0136EA 97.2 EUR
734-0136 734-0202 734-0109 734-0280 734-0141 734-0281 734-0275
Collagen I Zellkulturschalen, Corning® BioCoat™
Zellkulturschalen Zell- und Gewebekultur-Platten
PS, beschichtet mit Collagen Typ I (aus Rattenschwanz), steril, apyrogen.
  • Manufactured in a highly controlled environment and rigorously tested to assure product consistency and performance
  • Stable at room temperature when stored in dry conditions
Collagen I, found in most tissues and organs, is most plentiful in dermis, tendon and bone. It is an integral part of the framework that holds cells and tissues together and has been recognised as a useful matrix for improving cell culture. Used in vitro, collagen can exert effects on the adherence, morphology, growth, migration and differentiation of a variety of cell types.