Zellkultureinsätze, Falcon®
Lieferant: Corning
Falcon®
734-0060EA 385.78 EUR
734-0060 734-0038 734-0037 734-0036 734-0042 734-0053 734-0052 734-0041 734-0063 734-0062 734-0040 734-0051 734-0061 734-0035 734-0034 734-0397 734-0033 734-0032
Zellkultureinsätze, Falcon®
Mikrotiterplatten Einsetz-Platten und Einsätze
Track-etched PET membranes have a smooth surface and defined cylindrical pores that traverse the membrane.
  • Low protein binding PET membrane
  • Sterilized by gamma irradiation
  • A wide variety of configurations including 6, 12 and 24 well
  • A broad selection of membrane pore sizes, 0,4, 1,0, 3,0 and 8,0 µm diameter
  • Packed in individual blister packs, 48 inserts/case
  • Non-Tissue Culture-treated insert housings prevent promiscuous growth of cells on the insert walls
  • Innovative hanging design facilitates pipetting and allows for co-culture