One-Step SYBR® Green qRT-PCR, qScript™
Lieferant: Quantabio
733-2079EA 219 EUR
733-2079 733-2078 733-2077 733-2076 733-2075 733-2082
One-Step SYBR® Green qRT-PCR, qScript™
Nukleinsäure-Reagenzien qPCR/RT-qPCR Enzymes and Kits
Lieferumfang: Kits are available with or without ROX or with fluorescein for Bio-Rad iCycler iQ® systems. All kits contain qScript One-Step Reverse Transcriptase (optimised 50X formulation of recombinant MMLV reverse transcriptase for one-step RT-PCR); One-Step SYBR Green Master Mix (2X) reaction buffer containing dNTPs, MgCl?, AccuStart™ Taq DNA polymerase, stabilisers, and SYBR® Green I dye; and nuclease-free water.