Taq DNA-Polymerase
Lieferant: Avantor
VWR®
733-1819EA 501 EUR
733-1819 733-1823 733-1307 733-2009 733-1305 733-1302 733-1313 733-1820 733-1303 733-1300 733-1301 733-1312
Taq DNA-Polymerase
Nukleinsäure-Reagenzien Enzyme und Kits für Endpunkt-PCR
VWR® Taq DNA Polymerase is an ultra-pure, thermostable, recombinant DNA polymerase, which provides robust PCR performance in a wide range of PCR applications, without time-consuming optimisation. The enzyme is isolated from Thermus aquaticus, and has a molecular weight of approximately 94 kDa. VWR® Taq DNA Polymerase has both a 5' to 3' DNA polymerase and a double strand 5' to 3' exonuclease activity. It leaves an A overhang, which makes the enzyme ideal for TA cloning.
  • Ideal choice for routine applications
  • High performance, thermostable DNA polymerase
  • Optimal for TA cloning
Taq DNA polymerase concentration: 5 Units/μl
EU = Units
Lieferumfang: VWR® Taq DNA Polymerase is usually supplied with either or both Key Buffer and Extra Buffer. Key Buffer (NH⁴⁺) gives a superior amplification signal (high yield) minimising the need for optimisation of the Mg²⁺ concentration, or the annealing temperature in most primer-template systems. Extra Buffer is a traditional potassium (K⁺) buffer. Extra Buffer promotes high specificity, but careful optimisation of primer annealing temperatures and Mg²⁺ concentrations may be required.