Storage microplates 384-well polypropylene, Corning®
Lieferant: Corning
Corning®
732-5551EA 1048.39 EUR
732-5551 732-5550 736-0202 732-0483 732-5530 732-5529 732-5528
Storage microplates 384-well polypropylene, Corning®
Mikrotiterplatten Deep-Well-Mikroplatten
These 384-well polypropylene plates and blocks are built to the industry standard footprint.
Robolid combines the sealing ability of a storage mat with the rigidity of a plastic lid and is designed for repeated application and removal by automation and to prevent short-term evaporation. Robolid also features silicone sealing plugs for organic solvent resistance and low extractables.
They are automation-friendly and solvent-resistant. They feature round-, conical-, or v-bottoms for optimal liquid handling and a rounded square-well geometry for reduced wicking. Both plates and blocks are certified DNase- and RNase-free.
Bestellinformation: The 384-well plates are packaged 25 per pack, except for CORN3672 which is packaged 10 per pack. The 384-well blocks are packaged 5 per pack.