Brilliant III SYBR® MM, Agilent Technologies
Lieferant: Agilent Technologies
AGLS600887EA 5571 EUR
AGLS600887 732-3799 AGLS600883 AGLS600886
Brilliant III SYBR® MM, Agilent Technologies
Nukleinsäure-Reagenzien qPCR/RT-qPCR Enzymes and Kits
Brilliant III Ultra-Fast SYBR® QPCR and QRT-PCR Master Mixes deliver superior specificity by utilizing a novel faster-activating hot start method to minimize the formation of primer-dimers and other off-target reactions.
  • Reliable and reproducible data
  • Novel fast Taq mutant for qPCR results in under 35 minutes
  • Enhanced rapid hot start capability saves time and reduces primer-dimer formation
  • Convenient pre-blended formulations compatible with any SYBR Green dye
These unique reagents are equally robust and reproducible across a variety of assays on fast cycling real-time platforms.
Bitte beachten For research use only. Not for use in diagnostic procedures.