Cooling block incubators, N°ICE and BL°CKICE
Lieferant: Cole Parmer
732-1474EA 1364.89 EUR
732-1474 732-1473
Cooling block incubators, N°ICE and BL°CKICE
Blockthermostate Heiz-/Kühl-Blockthermostate
Maintains reagents at 4 °C for the whole working day
Autoclavable ceramic coated beads
Bestellinformation: BL°CKICE is supplied without block inserts, please order these separately. N°ICE is supplied complete with bucket and beads.