Lames micro-array, HTA™ Slides
Lieferant: Greiner bio-one
731-3014EA 162 EUR
731-3014 731-3021 731-3010 731-3013 731-3012
Lames micro-array, HTA™ Slides
Objektträger Mikroarray-Objektträger
The HTA™ 1 can be used for the processing of one sample. The HTA™ 12 are partitioned into 12 flat compartments, each with a printable surface of 6×6 mm and a low well rim of 0,5 mm. Twelve samples can be processed simultaneously.
  • Multifunctional 3-D amino, 3-D aldehyde or 3-D epoxy surface functionality
  • Low well rim of only 0,5 mm, low auto-fluorescence
  • Plastic plates for high-throughput microarrays
Combine the advantages of the universal microplate platform with microarrays for diagnostic applications due to identical well geometry compared with a standard 96-well microplate.