Blood gas capillary tubes, VITREX®
Lieferant: Vitrex Medical
MODU182613EA 113 EUR
MODU182613 720-2297 720-2298 720-2296 MODU182013 720-2299 MODU183313
Blood gas capillary tubes, VITREX®
Pipetten Mikrokapillaren
These disposable capillary tubes for blood gas analyses are made from heavy walled glass.
  • Available non-heparinised, with sodium or lithium heparinisation
  • Careful fire polishing, heavy walled tubes and robust end-caps secure proper function and reduce operating risk
  • Both ends melted
Shelf life: 4 years.