Trichterkammern, CytoSep™, für HETTICH Cyto-System
Lieferant: Simport Scientific
CytoSep™
720-1955EA 122 EUR
720-1955 720-1872 720-1873 720-1954
Trichterkammern, CytoSep™, für HETTICH Cyto-System
Trichter Cytologietrichter
The CytoSep™ funnel chambers for the Hettich Cyto-System optimises laboratory throughput with multiple funnel options in 2, 3, or 4 funnels per slide.
  • These funnels are ideal for applications such as CSF, viral infections, or bronchial secretions, and are available in four sizes from 1 to 8 ml. The 1 ml chamber is for small volume samples of low cell content. Both 2 and 4 ml chambers are suggested for cell-rich samples, e.g. pleura, ascites and bronchial washings, while the 8 ml chamber is perfect for large volume samples such as urine.