VWR®, Universale pH-Elektroden
Lieferant: VWR
VWR®
662-1382EA 89.6 EUR
662-1382 662-1393 662-1392 662-1391 662-1390 662-1397 662-1386 662-1396 662-1385 662-1384 662-1395 662-1383 662-1401 662-1389 662-1399 662-1388 662-1387 662-1398 662-1405 662-1404 662-1403 662-1402 662-1407 662-1406
VWR®, Universale pH-Elektroden
Elektroden pH-Elektroden
Laboratory and field electrodes for general applications. Supplied with a fixed cable or plug head, VWR® electrodes can be connected to most pH meters on the market.
All VWR® electrodes are checked at two points during the manufacturing process and then lot stamped to ensure quality. They are supplied with a soaker storage bottle and cap to make sure the pH-sensitive glass and reference junction remain in top condition, and are instantly ready for use.
Lieferumfang: Includes pH-electrode only.