Quarz-Kontrollplatten
Lieferant: Bellingham + Stanley
635-0405EA 1838 EUR
635-0405 635-0666 635-0023 BELL90-803 BELL90-805 635-0022
Quarz-Kontrollplatten
Standards Refraktometrie-Standards
Quartz control plates for the calibration and verification of polarimeters. of optical and digital polarimeters and saccharimeters.
  • Inert quartz with relatively low temperature coefficient, easy to handle, and infinite shelf life
  • Available in three optical rotation values
  • Three decimal place uncertainty
The optional temperature saddles may used as an interface between the quartz plate and instrument temperature sensor to provide fine compensation.
Standards: Traceable to PTB. UKAS-MRA calibration in accordance with ISO 17025:2005 between 325-880nm available.
Bestellinformation: Sadlles can be ordered in option.
Lieferumfang: Supplied with a certificate of verification in a plastic storage case.