Polarimeter tubes and spare parts
Lieferant: Bellingham + Stanley
BELL35-10EA 432 EUR
BELL35-10 BELL35-96 635-0029 BELL35-95 635-0027 BELL35-82 635-0028 635-0289 635-0025 635-1005 635-1004 635-0026 BELL35-91 BELL35-18 BELL35-17 635-0024 635-0030 BELL35-11 BELL35-88 635-0290 BELL35-90
Polarimeter tubes and spare parts
Polarimeter
Polarimeter tubes are manufactured to high quality standards where tube ends are precision ground and windows made from selected low strain glass providing highest accuracy along the light path.
  • Jacketed funnel flow-through tubes for sugar testing laboratories
  • Water-jacketed tubes can be used to provide temperature control as required
  • Continuous flow through tubes allows monitoring of pumped samples
Standards: Conforming to ICUMSA recommendations.
Bestellinformation: XPC adaptors and cover glasses for UV measurements are also available.