Temperatur-Indikatorstreifen
Lieferant: Testo
620-9125EA 35 EUR
620-9125 620-9126 620-9127 620-9122 620-9123 620-9124
Temperatur-Indikatorstreifen
Temperatur-Indikatorstreifen
These indicators are self-adhesive, temperature proof foils with temperature sensitive elements for temperature control and regulation. They are particularly suitable for monitoring temperature in small objects.
  • Change in colour within two seconds
  • Practical booklet with 10 indicators
  • Maximum storage temperature: 25 °C; storage in a refrigerator is recommended
Verpackung: Ab 5000 Stück (100 Blatt mit je 50 Stück).