pH food electrode
Lieferant: Testo
620-3662EA 229 EUR
620-3662
pH food electrode
Elektroden pH-Elektroden
Robust and dirt-insensitive pH penetration electrode without temperature sensor.
  • With hole diaphragm
  • Unbreakable plastic coating around glass electrode
  • Maintenance-free food-safe gel electrolyte
Ideal for use with Testo 230 instrument.