This luxmeter permits measurements of light intensity. It provides a sensor adjusted to the spectral sensitivity of the eye. A backlit display and an auto-off function complete the instrument.
- Hold-Funktion für ein komfortables Ablesen der Messwerte
- Max-/Min-Werte
- Betriebstemperatur von 0 bis +50 °C
The testo 540 lux meter can be used to measure both artificial and natural light and provides with a quick and easy means of measuring lighting conditions in the workplace. This meter is a pocket-sized entry level model designed specifically for carrying out spot checks in the workplace, shops and offices.
Lieferumfang: Inkl. Handschlaufe, Gürteltasche, Schutzkappe, Kalibrier-Protokoll und 2 Mignon Batterien (AAA) für ?20 h Betriebsdauer (ohne Display-Beleuchtung).