Hygroscopic materials always try to form a moisture equilibrium with the surrounding air. This principle is used to measure the moisture in material by measuring moisture equilibrium.
- Slim design (Ø 4 mm) with remote electronics
Ideal for tight measuring points, based on the moisture equilibrium measured, testo 635 automatically shows material moisture in % in the display.
Material moisture calculation is available for the following materials:
Concrete
High-insulation bricks
Solid bricks
Hard wood
Soft wood
Gas concrete
Particle board
Calcium sulphate floating screed
Cement floating screed
Lime sand brick
Materials are easy to select from the menu. Using the 'Material' user profile, the material can be accessed directly at the touch at a button.
Additional material characteristics can be stored in testo 635-2 using ComSoft.
Lieferumfang: Supplied with thin humidity probe (Ø 4 mm) with PTFE protective cap and adhesive modelling clay (four items), including fixed cable.