Graduated pipettes, Class AS
Lieferant: Witeg
613-2103EA 52.1 EUR
613-2103 613-2102
Graduated pipettes, Class AS
Pipetten Messpipetten
Graduated pipettes, class AS, conformity certified.
  • For complete swift delivery
  • With amber stain main point ring-graduation
  • Tempered delivery tips offering a wear resistance
  • With construction in the suction to hold cotton plug
  • Waiting time 3 s
Standards: DIN 12697