Büretten, gerader Hahn, DURAN®, Schellbach
Lieferant: DWK Life Sciences
612-5226EA 224.86 EUR
612-5226 612-5232 612-5230 612-5231 612-5227 612-5228
Büretten, gerader Hahn, DURAN®, Schellbach
Büretten
DURAN®, Borosilikatglas 3.3, klar, Klasse AS.
  • With Schellbach stripe and main graduations as circular divisions
  • Straight stopcock
  • With glass key
  • Marks and inscription in blue
  • Calibration is based on the poured out volume ("Ex")
Complies to the DIN ISO 385
With printed retrace code (batch identification).