Pasteur pipettes, Corning®
Lieferant: Corning
Corning®
612-5199EA 73 EUR
612-5199 612-5200 612-5198
Pasteur pipettes, Corning®
Pipetten Pasteur- und Transfer-Pipetten
Soda-lime or borosilicate glass.
  • Single use eliminates the danger of cross-contamination of valuable specimens or laboratory reagents
  • Features a constriction one inch below the top to facilitate plugging
  • Available in convenient bulk packs
These disposable Pasteur pipettes are ideal for rapid non volumetric transfer work in bacteriology, immunology, haematology and serology studies, as well as blood bank and clinical chemistry procedures.