Serologische Pipetten, PYREX®
Lieferant: Corning
PYREX®
612-5138EA 717.6 EUR
612-5138 612-5136 612-5133 612-5177 612-5134 612-5178 612-5750 612-5131 612-5132 612-5139 612-5130 612-5193 612-5194 612-5192 612-5188 612-5166 612-5189 612-5186 612-5164 612-5187 612-5165 612-5748 612-5749 612-5162 612-5140 612-5141 612-5163
Serologische Pipetten, PYREX®
Pipetten Serologische Pipetten
These sterile disposable 1 ml PYREX® serological pipettes, calibrated (TD) with blow-out, offer long, slender, tapered tips to make pipetting go faster and easier.
  • All pipettes have negative graduations
  • Fire-polished tips for burr-free, even flow rates
Easy-open Steri-View™ wrappers (one side paper, one side plastic) have color-coded sizes printed directly on the paper for easy identification by size.
Standards: Colors conform to I.S.O. standards.
Verpackung: Non sterile pipettes available in bulk package; sterile pipettes available in individual paper-plastic wrappers or as Multi-Pack in plastic bags.