Graduated pipettes, type 2, Class AS, total delivery
Lieferant: Hirschmann Laborgeräte
612-4157EA 10 EUR
612-4157 612-4156 612-4159 612-4158 612-4153 612-4155 612-4154 612-4711 HIRS1101329 612-4713
Graduated pipettes, type 2, Class AS, total delivery
Pipetten Messpipetten
AR-Glas®.
  • Calibrated to deliver (TD, Ex); Type 2: nominal volume on top
  • Marks and inscriptions in high contrast blue enamel
  • With standard cotton plug from five ml
  • Certificate of conformity with dated batch identification available on internet (www.hirschmannlab.de)
DIN EN ISO 835
Length: 360 mm ±5 mm except for 25 pipettes: 450 mm ±5 mm
Standards: DIN EN ISO 835. Conformity certified (DE-M marking).