Biocompatible in-line filters, Upchurch Scientific®
Lieferant: IDEX Health & Science
Upchurch Scientific®
UPCHA-428XEA 192 EUR
UPCHA-428X UPCHA-431 554-3039 554-3038
Biocompatible in-line filters, Upchurch Scientific®
Filter In-Line Filter
Inline filters consist of a stainless steel body and two PEEK end fittings.
  • Maximum recommended flow rate of 25 ml/min for A-430 and 10 ml/min for A-431
  • Designed to connect 1/16" OD or smaller tubing
  • Pressure rated to 6000 psi (414 bar)
  • 10-32 coned threads