In-line filters, Upchurch Scientific®
Lieferant: IDEX Health & Science
Upchurch Scientific®
UPCHA-313EA 173 EUR
UPCHA-313 554-3019 554-3018 554-1781
In-line filters, Upchurch Scientific®
Filter In-Line Filter
Placed between pump and sample injection valve to trap particles released through normal piston seal wear.
  • For 1/16" OD tubing
  • Pressure rated to 6000 psi (414 bar)