VWR® vial kit for Karl Fischer titration
Lieferant: VWR, part of Avantor
VWR®
548-2398EA 357 EUR
548-2398
VWR® vial kit for Karl Fischer titration
Vials Chromatographie-Vials
This vial kit is compatible with oven sample processors 774.
  • Tight and reliable vial/cap combination
  • Keep the water content constant, as well for longer transport/storage times