VWR® ND11, Rollrandflaschen
Lieferant: VWR
VWR®
548-0906EA 348 EUR
548-0906 548-3092 548-0171 548-8010 548-2159 548-2126 548-3005 548-0003 548-3004 548-0474 548-3003 548-0447 548-0004 548-3337 548-0005
VWR® ND11, Rollrandflaschen
Vials
Diese Flaschen sind für Instrumente der folgenden Hersteller geeignet: Agilent, Carlo Erba, CTC, DANI, Fisons, Gerstel, Jasco, PerkinElmer, Shimadzu, Spark, Thermo Scientific, Varian etc.
  • Standard vials for GC and HPLC
  • Vials with a barcode label, pre-crimped vials are also available
  • Microlitre vials for sample preparation (reactions, concentrations) or as an alternative for conical micro-vials or crimp neck vials with inserts
TopSert vials are an alternative to glass vials with fused-in micro-inserts or micro-insert with plastic spring. The TopSert glass micro-insert is centred in the plastic mould and pressed firmly against the septa due to its slightly overlapping edge.
Lieferumfang: Sets enthalten: 1,5 ml Rollrandflaschen (11,6×32 mm), Bördelkappe aus Aluminium, klar lackiert, mit zentralem Loch (5,5 mm) und Septum.