Scheidetrichter, konisch
Lieferant: Lenz
532-0105EA 216.72 EUR
532-0105 532-0104
Scheidetrichter, konisch
Trichter Scheidetrichter
Separating funnels, conical, made of borosilicate glass 3.3, amber coloured.
  • Amber coloured to protect light sensitive substances
  • Large labeling field
  • Very good chemical and temperature resistance
With interchangeable PTFE key.