Zentrifugenröhrchen
Lieferant: Greiner bio-one
391-3470EA 310 EUR
391-3470 525-0407 212-3166 525-0406 525-0405 212-3168 391-3476 391-3465
Zentrifugenröhrchen
Röhrchen Zentrifugenröhrchen
Transparent PS or PP, 2-position ventilation stopper, round bottom, sterile.
  • PS tubes without graduations, good thermal, mechanical and chemical resistance, centrifugation RFC 5000×g (fix-angle rotor) or 4800×g (swinging-bucket rotor)
  • PP tubes with graduations; high thermal, mechanical and chemical resistance, centrifugation RCF 34000×g (fix-angle rotor) or 5800×g (swinging-bucket rotor)
  • The ventilation stopper has two functions: when pushed in gently, it provides uniform ventilation of the tube contents; when pushed in firmly, it provides a tight seal
  • Free from DNase/RNase and human DNA, non-pyrogenic
Bestellinformation: Available in packs of 25 or wrapped individually.