Microsart® Manifolds accurately detect and quantify microorganisms in liquid samples. Simply select the proper manifold for your individual microbiological testing procedure, choose between reusable and single-use funnels or filtration units, and then decide between different sizes of manifolds and enjoy the manifold that adapts to your needs.
- 100% stainless steel for minimised cleaning effort and full autoclavability without taking apart
- Sterile venting in critical applications reduces risk of secondary contamination
- Quick connection adapters for fast connection of tubing, manifolds or end caps
- Rubber feet and low working height for safe and stable ergonomic working procedures
Microsart® Manifold for Microsart®@filter and Microsart Funnels (pharmaceutical applications, cosmetics: Manifold for single use filtration units)
• Add single sterile consumables: Microsart®@filter or Microsart Funnels
• For minimal risk of cross contamination by integrated venting and by touch-free membrane transfer when combining Microsart@filter use with Microsart@media plates
Microsart® Manifold for Biosart® 100 monitors (beverage and environmental water testing: Manifold for single-use filtration units)
• Add consumables: Biosart® 100 Monitor and Biosart® 100 media
• For minimised risk of cross contamination by integrated venting and by touch-free inoculation
• Media conforms to international regulations
Microsart® Manifold for Biosart® 250 funnels (optional beverage and environmental: Manifold for autoclavable funnels)
• Add autoclavable Biosart® 250 Funnels and Membrane filter
• Flexible easy quick connect set-up to adapt to variable numbers of tests per day
• Easy cleaning procedure, fully autoclavable manifold, no material mix
Microsart® Manifold with stainless steel funnels (optional beverage and environmental: Manifold with stainless steel funnels)
• Add membrane filter
• Flexible easy quick connect set-up to adapt to variable numbers of tests per day
• Easy cleaning by no dismantling, fully autoclavable manifold, no material mix
• Flammable stainless steel funnels (100 or 500 ml)
Lieferumfang: Manifolds include filter base support with sinter frit, blind coupling, 20 cm tubing and venting filter.