Vacuum membrane filter holder, Whatman™
Lieferant: Whatman products (Cytiva)
Whatman™
511-0137EA 516 EUR
511-0137 512-0641 516-0616 516-0614 516-0615 516-0613
Vacuum membrane filter holder, Whatman™
Trichter Membranfiltertrichter
Produced from borosilicate glass and available with a choice of support screen.
  • Sintered glass support is recommended for filtration and biological analysis
  • 304 stainless steel support screen is suitable for use with proteinaceous solutions
Suitable for aqueous and organic solvent filtration. The funnel seal ensures that the sample does not bypass the membrane and that particulates are retained on the surface of the membrane.
Chemical resistant to most aqueous and organic solutions as well as acid and caustic solution resistant.
Applications include: - food & beverage (e.g cold sludge in beer; ice cream), pharmaceuticals and cosmetics, water and wastewater, residue analytics and precipitation analysis, contamination tests (e.g in electroplating), microbiological, biochemical, and hydrobiological detection, radiochemical tests, particle analysis in sensitive areas of electronics, aviation, and space travel.