Spritzenvorsatz-/Inline-Filter, Nalgene®
Lieferant: Thermo Fisher Scientific
Nalgene®
513-2221EA 153 EUR
513-2221 513-2222
Spritzenvorsatz-/Inline-Filter, Nalgene®
Filter Spritzenvorsatzfilter
These syringe filters with PTFE membrane and PP housing are used for benchtop fermenters, in-line gas filtration for sterile air or CO₂ and to protect vacuum pumps from damage by aqueous solutions. They are also designed for filtration of aggressive chemicals and solvents such as those used in GC and HPLC.
Inlet/outlet: Stepped hose barb accepts six to nine mm Int.Ø. tubing.