Tissue vial box, blue, WHEATON® CryoFILE
Lieferant: DWK Life Sciences
WHEATON®
479-2114EA 222 EUR
479-2114 479-2113 479-2116 479-2115 479-2117 479-2112
Tissue vial box, blue, WHEATON® CryoFILE
Boxen Boxen für Vials
The CryoFILE tissue vial storage Box is used for batching and storing CryoELITE tissue vials.
CryoFILE partitions are numbered from 1 to 25 for easy content identification, with the numbering system printed on lid and bottom of box. CryoFILE tissue storage boxes are available in six colors, to provide easy sample identification. Made of water repellent material for longer durability, the boxes are able to withstand cryogenic temperatures to –196 °C.