Dri-block® Heaters, DB Range

Lieferant: Cole Parmer

Techne® Dri-block®
460-0058EA 1137.22 EUR
460-0058 460-0057 460-0054
Dri-block® Heaters, DB Range
Blockthermostate Blockthermostate, analog
The Techne® Dri-Blocks® are particularly suitable for microbiology and clinical laboratories for incubation, boiling, inactivation, wet ashing, sample concentration and enzyme analysis. There is a choice of temperature ranges; 25 to 100 °C or 25 to 200 °C. These models have a rotary knob for temperature setting (but no digital display).

  • Wide range of interchangeable blocks to accommodate tubes, microcentrifuge tubes and 96-well plates
  • Very accurate temperature control
  • Temperature stability at 40 °C is ±0,05 or ±0,1 °C, maximum temperature variation between identical blocks is ±0,2 °C
  • Rapid heat up rate: 8 minutes from 30 to 37 °C

Standards: IP Protection Class according to DIN EN 60529: IP 31.

Lieferumfang: Supplied with block extraction tool. Blocks must be ordered separately.

Bitte beachten Note: Temp. stability value shown above is at 100 °C.
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR